Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benchmark Scientific Digital Dry Bath Block Products for Science Education

Click to view available options
Tube Size:
0.2mL
0.5mL
1.5mL
10mL
15mL
No. of Places:
1
12
24
48

The Fisherbrand high-grade aluminum blocks are precision machined to match the conical size and shape of 0.5, 1.5, 2.0, 15 and 50mL tubes - providing unsurpassed temperature uniformity.

These high-grade aluminum blocks are precision machined to match the conical size and shape of 0.5, 1.5, 2.0, 15 and 50mL tubes - providing unsurpassed temperature uniformity.

Order Info

These high-grade aluminum blocks are precision machined to match the conical size and shape of 0.5, 1.5, 2.0, 15 and 50mL tubes - providing unsurpassed temperature uniformity.

Certifications

CE

Dimensions (L x W x H) 7.62 x 9.53 x 5.08 cm (3 x 3.75 x 2 in.)
Material Aluminum
For Use With (Equipment) Single-Block Digital Dry Baths
For Use With (Application) Heating and incubating samples
Holds 96 well PCR Plate
Tube Size 0.2mL
Warranty 2 Years
Type Accessory, Block
No. of Places 1