Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Digital Heated Ultrasonic Cleaner PROMO
SDP

Easy to use with dependable digital control panel. The 40 kHz ultrasonic bath cleans a broad range of materials, and the tank is constructed of stainless steel to resist corrosion and is easy to clean.

Catalog No. FB1533741PL


Explore available promotions
Add to Cart

Description

Description

  • Programmable from 1 to 99 minutes
  • Stainless steel tank
  • Basket and cover included
  • 2-year warranty
  • Optional use of heating to 80°C to break down debris and oils
  • 40 kHz ultrasonic frequency
  • High and low cleaning power settings
  • Degas mode
Specifications

Specifications

5 A
Digital
40 kHz
3 L
Basket and cover
2 Years
1 to 99 min.
Stainless Steel
120 V
240 x 140 x 100 mm (9.5 x 5.5 x 4 in.)
20°C to 80°C
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.