Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Argos Technologies Digital Mini Dispensing Pump Systems

Compact digital pump system boosts efficiency and ergonomics in media dispensing and filling applications

Manufacturer:  Argos Technologies AR7792230

 View more versions of this product

Catalog No. 03-391-221


Add to cart

Includes: Masterflex L/S 25 BioPharm pump tubing

Description

Description

Features:

  • Ideal for dispensing and filling culture media, buffer solutions, dilutents, nutrients, additives, and other fluids
  • Unlike hand pouring, manual pipetting, or bottletop style dispensers, these pumps can easily repeat dispense volumes with copy dispense function
  • Ideal for microbiology, tissue- and cell-culture labs filling plates, tubes, vials, flasks, or bottles
  • Programmable delay interval pauses pump between cycles
  • Durable ABS housing that is easy to wipe down and clean
  • Pump head features a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing
  • Self-lubricating thermoplastic polyester rollers are low friction and long wearing for smooth operation and long life
  • Programmable delay interval pump between cycles
  • Compact size makes it easy to move (20% smaller than competitive units)
  • Open-head sensor stops the drive when the pump head is opened
  • Pump head acceps continuous lengths of tubing for clean flow paths
  • BioPharm Plus tubing offers low spallation, long pumping life, and exceptional flow stability over time, making it ideal for accurate filling and dispensing applications. 
  • Automatic tubing retention makes tube loading quick and easy
  • Dispense volume: 0.1 to 9999mL
  • Remote control: Speed, direction, start, stop

 

Features:

  • Ideal for dispensing and filling culture media, buffer solutions, dilutents, nutrients, additives, and other fluids
  • Unlike hand pouring, manual pipetting, or bottletop style dispensers, these pumps can easily repeat dispense volumes with copy dispense function
  • Ideal for microbiology, tissue- and cell-culture labs filling plates, tubes, vials, flasks, or bottles
  • Programmable delay interval pauses pump between cycles
  • Durable ABS housing that is easy to wipe down and clean
  • Pump head features a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing
  • Self-lubricating thermoplastic polyester rollers are low friction and long wearing for smooth operation and long life
  • Programmable delay interval pump between cycles
  • Compact size makes it easy to move (20% smaller than competitive units)
  • Open-head sensor stops the drive when the pump head is opened
  • Pump head acceps continuous lengths of tubing for clean flow paths
  • BioPharm Plus tubing offers low spallation, long pumping life, and exceptional flow stability over time, making it ideal for accurate filling and dispensing applications. 
  • Automatic tubing retention makes tube loading quick and easy
  • Dispense volume: 0.1 to 9999mL
  • Remote control: Speed, direction, start, stop
Specifications

Specifications

0.11 to 315mL/min
0.1 to 300rpm
Yes
90 to 260VAC
50/60Hz
1
(repeatability) ±0.1%
Foot switch with dispensing stand, rod, clamp and precision stainless steel nozzle
IP33
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit