Learn More
Cole-Parmer Shimpo DT-725 Digital Stroboscope with Internal Battery; 115 VAC  DFS Item
Manufacturer:  Cole-Parmer 5356028
These light weight stroboscopes are designed for quality control, motion analysis, vibration problems, maintenance work, experiments in engineering school projects, production line checks, examination and speed measurement of motors under various loads, gear movement and engagement, alignment of parts, cams, propellers, fan blades, centrifuges, spindles, turbines and other machinery exhibiting repetitive motion. They also provide pertinent information on the motion of vibrating parts, oil leaks, liquid spray patterns, and a host of other industrial and scientific applications. The stroboscopes also feature phase shifting, rotary pulse indicator, floating decimal point and LED digital display, slave functions, and manual flash rate control. As electronic digital tachometers, you can measure RPM without contact or special markings.
Catalog No. 50-993-593