Learn More
GSC Digital Voltmeter
GSC Digital Voltmeters measure the potential difference, or voltage (V), between two points in a circuit.
Manufacturer:  GSC DVM01
 View more versions of this product
Catalog No. S38999
Add to cart
Specifications
Specifications
13.75 x 17.5 x 11.25cm
4.5in.
5.5in.
1.999kg
199.9uA
1
digital
11.25
13.75
17.5
Education Electricity
199.9mV