Learn More
Ansell™ Digitcare™ ICU 911-T™ Medical Exam Gloves
Ansell™ Digitcare™ ICU 911-T™ Medical Exam Gloves are preferred by America's First Responders in high-risk applications
Manufacturer:  Ansell™ DCER2B
 View more versions of this product
Catalog No. 19-500-984
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
  • Thick design for added strength and durability
  • Extended cuff for expanded protection over the wrist and forearm
  • Comfortable latex material for soft, elastic fit and feel
  • Processed so that it has an extremely smooth, and clean feeling surface for a unique touch
  • Fentanyl Permeation Testing: ICU911-T™ has zero breakthrough with Fentanyl Citrate 100 mcg/2 mL for 240 minutes (test limit) per ASTM D697
Specifications
Specifications
Yes
Extended
Box
100/Pk.
11 in.
20 mil
Latex
Medium
Cream
ASTM D3578, FDA21 CFR 177-2600, ISO 13485