Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diglycidyl ether, 95%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2238-07-5
C6H10O3
130.14
MFCD00080479
GYZLOYUZLJXAJU-UHFFFAOYNA-N
2-{[(oxiran-2-yl)methoxy]methyl}oxirane
C(OCC1CO1)C1CO1
2238-07-5
C6H10O3
130.14
MFCD00080479
GYZLOYUZLJXAJU-UHFFFAOYNA-N
2-{[(oxiran-2-yl)methoxy]methyl}oxirane
C(OCC1CO1)C1CO1
Boiling Point 260°C
Specific Gravity 1.12
Flash Point 64°C
Density 1.1200g/mL
Refractive Index 1.4460 to 1.448
Color Colorless to Brown
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Formula Weight 130.14
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Diglycidyl ether
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
May form explosive peroxides.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with

GHS Signal Word: Danger

EINECSNumber : 218-802-6