Alfa Aesar™ Diglycolic acid, 98%

Manufacturer: Alfa Aesar™ B2375822

Quantity 100g

 View more versions of this product

Catalog No. AAB2375822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 110-99-6
    Molecular Formula C4H6O5
    Formula Weight 134.09
    MDL Number MFCD00004309
    Beilstein 1072117
    Synonym 2,2'-Oxydiacetic acid
    Chemical Name or Material Diglycolic acid

    Specifications

    Formula Weight 134.09
    Melting Point 140° to 145°C
    Quantity 100g
    MDL Number MFCD00004309
    Synonym 2,2'-Oxydiacetic acid
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Molecular Formula C4H6O5
    CAS 110-99-6
    Beilstein 1072117
    RTECS Number AJ1530000
    Chemical Name or Material Diglycolic acid
    Assay 98%

    Description

    Applications
    Diglycolic acid was used in the preparation of needle-like aragonite particles from calcium nitrate aqueous solutions. It is also used in the synthesis of N-interlinked imipramines which show apoptotic activity against malignant cells as well as signs of combating Burkitt’s lymphoma.

    Solubility
    Soluble in water, methanol.

    Notes
    Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit