Learn More
Digoxigenin Mouse anti-All, Janelia Fluor 549, Clone: 21H8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP231191JF549
Catalog No. NBP23191549
Add to cart
Description
Description
Digoxigenin Monoclonal antibody specifically detects Digoxigenin in All samples. It is validated for Immunohistochemistry.
Specifications
Specifications
Digoxigenin
21H8
Immunohistochemistry
Purified
Mouse
IgG1 Kappa
0.1mL
Primary
Reacts with free and bound digoxigenin
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
DIG
Digoxigenin
Protein A purified
Store at 4°C in the dark.
Monoclonal
All