Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Digoxin, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Digoxin is used in several cardiac pathologies such as atrial fibrillation and heart failure.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Digoxin is used in several cardiac pathologies such as atrial fibrillation and heart failure.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Digoxin is used in several cardiac pathologies such as atrial fibrillation and heart failure.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Melting Point | ∼250°C (decomposition) |
Quantity | 1 g |
UN Number | UN2811 |
Merck Index | 14,3167 |
Solubility Information | Soluble in dimethyl sulfoxide and methanol. |
Formula Weight | 780.95 |
Percent Purity | 96% |
Chemical Name or Material | Digoxin |
Safety and Handling
Hazard Category | H300-H336-H351-H373 |
Hazard Statement | GHS H Statement H300 Fatal if swallowed. |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P330-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
EINECSNumber | 244-068-1 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction