Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydroergocristine mesylate, Tocris Bioscience™

Catalog Number 0474100
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
24730-10-7
C36H45N5O8S
707.843
SPXACGZWWVIDGR-SPZWACKZSA-N
dihydroergocristine mesylate, dihydroergocristinmesilat, unii-ds7cl18uam, dihydroergocristine mesilate, 9,10-dihydroergocristine methanesulfonate, ds7cl18uam, dihydroergocristine methanesulfonate, dihydroergocristine methanesulfonate salt, 5'alpha-benzyl-9,10alpha-dihydro-12'-hydroxy-2'-isopropylergotaman-3',6',18-trione monomethanesulphonate, diertine tn
444034
CHEBI:31490
CC(C)C1(C(=O)N2C(C(=O)N3CCCC3C2(O1)O)CC4=CC=CC=C4)NC(=O)C5CC6C(CC7=CNC8=CC=CC6=C78)N(C5)C.CS(=O)(=O)O
This item is not returnable. View return policy
24730-10-7
C36H45N5O8S
707.843
SPXACGZWWVIDGR-SPZWACKZSA-N
dihydroergocristine mesylate, dihydroergocristinmesilat, unii-ds7cl18uam, dihydroergocristine mesilate, 9,10-dihydroergocristine methanesulfonate, ds7cl18uam, dihydroergocristine methanesulfonate, dihydroergocristine methanesulfonate salt, 5'alpha-benzyl-9,10alpha-dihydro-12'-hydroxy-2'-isopropylergotaman-3',6',18-trione monomethanesulphonate, diertine tn
444034
CHEBI:31490
CC(C)C1(C(=O)N2C(C(=O)N3CCCC3C2(O1)O)CC4=CC=CC=C4)NC(=O)C5CC6C(CC7=CNC8=CC=CC6=C78)N(C5)C.CS(=O)(=O)O

Partial α agonist. Non-selective

5-HT receptor antagonist; partial agonist at adrenergic and dopaminergic receptors.
Quantity 100 mg
Formula Weight 707.84
Percent Purity >98%
Chemical Name or Material Dihydroergocristine mesylate
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit