Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydroergotamine Mesylate, USP, 97-103%, Spectrum™

Click to view available options
Quantity:
250 mg
1 g
5 g
100 g
Packaging:
Amber Glass Bottle
6190-39-2
C34H41N5O8S
679.79
ADYPXRFPBQGGAH-UMYZUSPBSA-N
(2R,4R,7R)-N-[(1S,2S,4R,7S)-7-benzyl-2-hydroxy-4-methyl-5,8-dioxo-3-oxa-6,9-diazatricyclo[7.3.0.0²,⁶]dodecan-4-yl]-6-methyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),9,12,14-tetraene-4-carboxamide; methanesulfonic acid
CS(O)(=O)=O.CN1C[C@@H](C[C@H]2[C@H]1CC1=CNC3=CC=CC2=C13)C(=O)N[C@]1(C)O[C@@]2(O)[C@@H]3CCCN3C(=O)[C@H](CC3=CC=CC=C3)N2C1=O
6190-39-2
C34H41N5O8S
679.79
ADYPXRFPBQGGAH-UMYZUSPBSA-N
(2R,4R,7R)-N-[(1S,2S,4R,7S)-7-benzyl-2-hydroxy-4-methyl-5,8-dioxo-3-oxa-6,9-diazatricyclo[7.3.0.0²,⁶]dodecan-4-yl]-6-methyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),9,12,14-tetraene-4-carboxamide; methanesulfonic acid
CS(O)(=O)=O.CN1C[C@@H](C[C@H]2[C@H]1CC1=CNC3=CC=CC2=C13)C(=O)N[C@]1(C)O[C@@]2(O)[C@@H]3CCCN3C(=O)[C@H](CC3=CC=CC=C3)N2C1=O

D1165, 6190-39-2, C33H37N5O5•CH4O3S

Spectrum™ Dihydroergotamine Mesylate, USP is an ergot alkaloid medication used to treat active migraine headaches. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Loss on Drying 0.04
Specific Rotation -1°6.7° to -22.7°
Quantity 250 mg
CAS Max % 1
Percent Purity 97 to 103%