Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dihydroergotoxine mesylate, Tocris Bioscience™
Click to view available options
Quantity:
100 mg
Description
A complex of closely related alkaloid salts. Binds with high affinity to the GABAA receptor Cl- channel, producing an allosteric interaction with the benzodiazepine site. Also interacts with central dopaminergic, serotonergic and adrenergic (α1) receptors. Displays antiproliferative activity in vitro (IC50 = 18 - 38 μM in prostate cancer cells) and exhibits cognition-enhancing, anticonvulsant and sedative activity in vivo. Orally active.
Specifications
Quantity | 100 mg |
Percent Purity | >98% |
Chemical Name or Material | Dihydroergotoxine mesylate |
Safety and Handling
Recommended Storage : Store at Room Temperature
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title