Dihydrogen dinitrosulfatoplatinate(II) solution, Pt 4-6% (cont. Pt), Alfa Aesar™

Manufacturer: Alfa Aesar 12627NA

Quantity ≈2g

 View more versions of this product

Catalog No. AA12627NA

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12033-81-7
    Physical Form Liquid
    Molecular Formula H2Pt(NO2)2SO4
    Formula Weight 384.17 (anhydrous)
    MDL Number MFCD00064668
    Synonym Dinitrosulfatoplatinum(II)
    Chemical Name or Material Dihydrogen dinitrosulfatoplatinate(II) solution, Pt 4-6% (cont. Pt)

    Specifications

    Formula Weight 384.17 (anhydrous)
    Quantity ≈2g
    Color Brown
    DOT Information DOT Class: 8, Packaging Group: III
    Physical Form Liquid
    Molecular Formula H2Pt(NO2)2SO4
    Appearance Brown liquid
    MDL Number MFCD00064668
    CAS 12033-81-7
    Synonym Dinitrosulfatoplatinum(II)
    Health Hazard 1 H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.
    Chemical Name or Material Dihydrogen dinitrosulfatoplatinate(II) solution, Pt 4-6% (cont. Pt)

    H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit