Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dihydrogen hexabromoplatinate(IV) nonahydrate, 99.9% (metals basis), Pt 23.7% min

Manufacturer:  Alfa Aesar™ 1261703

Quantity 1g

 View more versions of this product

Catalog No. AA1261703


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

20596-34-3
838.67 (676.56 Anhydrous)
UN3260
Dihydrogen hexabromoplatinate(IV) nonahydrate
H2PtBr6·9H2O
MFCD00049645
Bromoplatinic acid; Platinic bromide

Specifications

Pt 23.7% min.
20596-34-3
H2PtBr6·9H2O
838.67 (676.56 Anhydrous)
UN3260
Dihydrogen hexabromoplatinate(IV) nonahydrate
1g
Crystalline
Red-brown
MFCD00049645
Bromoplatinic acid; Platinic bromide
99.9%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H334-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P284a-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit