Alfa Aesar™ Dihydrogen hexachloroosmate(IV) hydrate, Premion™, 99.95% (metals basis), Os 37% min

Manufacturer: Alfa Aesar™ 1217803

Quantity 1g

 View more versions of this product

Catalog No. AA1217803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 27057-71-2
    Linear Formula H2OsCl6·xH2O
    Formula Weight 404.93 (Anhydrous)
    MDL Number MFCD00011329
    UN Number UN3260
    Synonym Hydrogen hexachloroosmate(IV); Chloroosmic acid
    Chemical Name or Material Dihydrogen hexachloroosmate(IV) hydrate
    Grade Premion™

    Specifications

    Formula Weight 404.93 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Physical Form Crystalline
    Color Black
    MDL Number MFCD00011329
    CAS 27057-71-2
    UN Number UN3260
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    Synonym Hydrogen hexachloroosmate(IV); Chloroosmic acid
    Health Hazard 1 Danger
    Sensitivity Hygroscopic
    Quantity 1g
    Linear Formula H2OsCl6·xH2O
    Concentration Os 37% min.
    Chemical Name or Material Dihydrogen hexachloroosmate(IV) hydrate
    Assay 99.95%, (metals basis)
    Grade Premion™

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit