Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dihydrogen hexachloroplatinate(IV) hexahydrate, low nitrate, Premion™, 99.95% (metals basis), Pt 37.5% min

Manufacturer:  Alfa Aesar™ 4281506

Quantity 5g

 View more versions of this product

Catalog No. AA4281506


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

18497-13-7
517.91 (409.82 Anhydrous)
UN2507
Dihydrogen hexachloroplatinate (IV) hexahydrate, Low nitrate
H2PtCl6·6H2O
MFCD00149910
14,7526
Premion™

Specifications

Pt 37.5% min
5g
Odorless
2.430g/mL
Red-brown
MFCD00149910
UN2507
Dihydrogen hexachloroplatinate (IV) hexahydrate, Low nitrate
Premion™
Hygroscopic
18497-13-7
Lumps
H2PtCl6·6H2O
517.91 (409.82 Anhydrous)
60°C (dec.)
14,7526
99.95%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300-H334-H372-H314-H318-H290-H317
Fatal if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
May cause an allergic skin reaction.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CHLOROPLATINIC ACID, SOLID

RTECSNumber : TP1500000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit