Dihydrogen hexachloroplatinate(IV) hydrate, Premion, 99.999% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 4369603

Quantity 1g

 View more versions of this product

Catalog No. AA4369603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 26023-84-7
    Physical Form Crystals
    Assay Percent Range 99.999% (metals basis)
    Molecular Formula H2PtCl6·xH2O
    Formula Weight 409.82
    MDL Number MFCD00149909
    Synonym Hexachloroplatinic acid hydrate
    Chemical Name or Material Dihydrogen hexachloroplatinate(IV) hydrate

    Specifications

    Physical Form Crystals
    Formula Weight 409.82
    Quantity 1g
    Color Red-brown
    DOT Information DOT Class: 8, Packaging Group: III
    Health Hazard 2 Do not breathe dust. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
    EINECS Number 241-010-7
    Odor Odorless
    Assay Percent Range 99.999% (metals basis)
    MDL Number MFCD00149909
    CAS 26023-84-7
    Synonym Hexachloroplatinic acid hydrate
    Health Hazard 1 H301-H317-H334-H314-H318-H290-H400-H410: Toxic if swallowed. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. May be corrosive to metals. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Molecular Formula H2PtCl6·xH2O
    Chemical Name or Material Dihydrogen hexachloroplatinate(IV) hydrate

    Do not breathe dust. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

    H301-H317-H334-H314-H318-H290-H400-H410: Toxic if swallowed. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. May be corrosive to metals. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit