Alfa Aesar™ Dihydrogen hexafluorozirconate, 20% w/w aq. soln.

Manufacturer: Alfa Aesar™ 39461A7

Quantity 5kg

 View more versions of this product

Catalog No. AA39461A7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12021-95-3
    Formula Weight 207.22
    MDL Number MFCD00082965
    UN Number UN3264
    Chemical Name or Material Dihydrogen hexafluorozirconate

    Specifications

    Formula Weight 207.22
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Color Pale green
    UN Number UN3264
    Physical Form Liquid
    MDL Number MFCD00082965
    CAS 12021-95-3
    Quantity 5kg
    TSCA TSCA
    Density 1.52g/mL
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H302-H312-H332
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
    Concentration 20% w/w aq. soln.
    Chemical Name or Material Dihydrogen hexafluorozirconate

    GHS H Statement
    H314-H302-H312-H332
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit