Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dihydrogen hexahydroxyplatinate(IV), 99.9% (metals basis), Pt 61.0% min

Manufacturer:  Alfa Aesar™ 1105203

Quantity 1g

 View more versions of this product

Catalog No. AA1105203


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

51850-20-5
MFCD00058744
Dihydrogen hexahydroxyplatinate(IV)
299.17
Hexahydroxyplatinic acid; Hydrogen hexahydroxyplatinate(IV)

Specifications

Pt ≤61.0%
1g
Crystalline
299.17
Hexahydroxyplatinic acid; Hydrogen hexahydroxyplatinate(IV)
99.9%, (metals basis)
Hygroscopic
51850-20-5
Pale to dark yellow
MFCD00058744
Dihydrogen hexahydroxyplatinate(IV)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit