Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydrokainic acid, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
1mg
50mg

Chemical Identifiers

52497-36-6
C10H17NO4
215.249
JQPDCKOQOOQUSC-OOZYFLPDSA-N
(2S, 3S, 4R)-2-Carboxy-4-isopropyl-3-pyrrolidineacetic acid
107883
CHEBI:43562

EAAT2 (GLT-1)-selective non-transportable inhibitor of L-glutamate and L-aspartate uptake

EAAT2(GLT1)-selective non-transportable inhibitor of L-glutamate and L-aspartate uptake (Ki = 23 μM). 130-fold selective over EAAT1 and EAAT3 (Ki > 3 mM). Also available as part of the Excitatory Amino Acid Transporter Inhibitor Tocriset™ .

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
CAS 52497-36-6
Molecular Formula C10H17NO4
Synonym (2S, 3S, 4R)-2-Carboxy-4-isopropyl-3-pyrrolidineacetic acid
InChI Key JQPDCKOQOOQUSC-OOZYFLPDSA-N
SMILES CC(C)C1CNC(C1CC(=O)O)C(=O)O
IUPAC Name (2S,3S,4R)-3-(carboxymethyl)-4-propan-2-ylpyrrolidine-2-carboxylic acid
Molecular Weight (g/mol) 215.249
PubChem CID 107883
ChEBI CHEBI:43562
Formula Weight 215.25
Chemical Name or Material Dihydrokainic acid
Grade >98%
Show More Show Less

Recommended Storage : Store at Room Temperature