Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dihydrolipoamide Dehydrogenase/DLD Antibody (SR2009), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Dihydrolipoamide Dehydrogenase/DLD |
---|---|
Clone | SR2009 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
Dihydrolipoamide Dehydrogenase/DLD Monoclonal antibody specifically detects Dihydrolipoamide Dehydrogenase/DLD in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
Dihydrolipoamide Dehydrogenase/DLD | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
2-oxo-glutarate complex, branched chain keto acid dehydrogenase complex), branched chain keto acid dehydrogenase complex, dihydrolipoamide dehydrogenase (E3 component of pyruvate dehydrogenase complex, dihydrolipoamide dehydrogenasemitochondrial, DLDH, E3, E3 component of pyruvate dehydrogenase complex, 2-oxo-glutarate complex, EC 1.8.1, EC 1.8.1.4, GCSL, Glycine cleavage system L protein, glycine cleavage system protein L, lipoamide dehydrogenase, lipoamide reductase, lipoyl dehydrogenase | |
A synthesized peptide derived from human Dihydrolipoamide Dehydrogenase/DLD (Uniprot #: P09622) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2009 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cellular Markers, Diabetes Research, Mitochondrial Markers, Neuroscience | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
1738 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title