Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydrolipoamide Dehydrogenase/DLD Mouse anti-Human, HRP, Clone: OTI6D5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270574H

Catalog No. NBP270574H


Add to cart

Description

Description

Dihydrolipoamide Dehydrogenase/DLD Monoclonal antibody specifically detects Dihydrolipoamide Dehydrogenase/DLD in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Dihydrolipoamide Dehydrogenase/DLD
OTI6D5
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
DLD
Full length recombinant protein of human Dihydrolipoamide Dehydrogenase/DLD (NP_000099) produced in HEK293T cells.
Immunogen affinity purified
Cellular Markers, Diabetes Research, Mitochondrial Markers, Neuroscience
Primary
1738.0
Immunohistochemistry, Western Blot
HRP
PBS with No Preservative
2-oxo-glutarate complex, branched chain keto acid dehydrogenase complex), branched chain keto acid dehydrogenase complex, dihydrolipoamide dehydrogenase (E3 component of pyruvate dehydrogenase complex, dihydrolipoamide dehydrogenasemitochondrial, DLDH, E3, E3 component of pyruvate dehydrogenase complex, 2-oxo-glutarate complex, EC 1.8.1, EC 1.8.1.4, GCSL, Glycine cleavage system L protein, glycine cleavage system protein L, lipoamide dehydrogenase, lipoamide reductase, lipoyl dehydrogenase
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit