Learn More
Selleck Chemical LLC Dihydroxy phenylglycine-25MG
SDP
Supplier:  Selleck Chemical LLC E125725MG
Encompass_Preferred
(R S)-3 5-DHPG hydrochloride (DHPG Dihydroxy phenylglycine) is a salt of free amino acid dihydroxy phenylglycine (DHPG) which acts as a selective and potent agonist of group I metabotropic glutamate receptors (mGluR) (mGluR 1 and mGluR 5)
Catalog No. 50-313-7252