Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydroxyaluminum Aminoacetate, USP, 94-102%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
41354-48-7
41354-48-7

D1057, 41354-48-7, C2H6AlNO4•xH2O

Spectrum™ Dihydroxyaluminum Aminoacetate, USP is used primarily as an antacid. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Pail
Loss on Drying 0.145
Quantity 2.5 kg
CAS Max % 1
Percent Purity 94 to 102%