Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dihydroxydiphenylsilane, 95%

Manufacturer:  Alfa Aesar™ 6915114

Quantity 25g

 View more versions of this product

Catalog No. AA6915114

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

947-42-2
216.32
UN1325
Dihydroxydiphenylsilane
(C6H5)2Si(OH)2
MFCD00002101
Diphenylsilanediol

Specifications

947-42-2
(C6H5)2Si(OH)2
MFCD00002101
Diphenylsilanediol
25g
Dihydroxydiphenylsilane
Powder
216.32
UN1325
138° to 142°C (dec.)
53°C (127°F)
95%

Safety and Handling

GHS H Statement
H228

GHS P Statement
P210-P280-P240-P241-P370+P378a

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit