Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diiodo[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II), Thermo Scientific™

Click to view available options
Quantity:
1 g
MFCD14155730
PdI2(dtbpf); [1,1'-Bis(di-tert-butylphosphino)ferrocene]diiodopalladium(II)
MFCD14155730
PdI2(dtbpf); [1,1'-Bis(di-tert-butylphosphino)ferrocene]diiodopalladium(II)

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Color Brown
Quantity 1 g
Sensitivity Air and light sensitive
Formula Weight 834.66
Physical Form Crystalline Solid
Chemical Name or Material Diiodo[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II)

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen