Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diisobutyl adipate, 97%

Click to view available options
Quantity:
25g
100g
500g
141-04-8
C14H26O4
258.358
MFCD00053722
RDOFJDLLWVCMRU-UHFFFAOYSA-N
diisobutyl adipate,isobutyl adipate,diba,ftaflex diba,plasthall diba,bis 2-methylpropyl hexanedioate,hexanedioic acid, bis 2-methylpropyl ester,adipic acid, diisobutyl ester,diisobutyl hexanedioate,adipic acid bis 2-methylpropyl ester
8831
CHEBI:34708
bis(2-methylpropyl) hexanedioate
CC(C)COC(=O)CCCCC(=O)OCC(C)C
141-04-8
C14H26O4
258.358
MFCD00053722
RDOFJDLLWVCMRU-UHFFFAOYSA-N
diisobutyl adipate,isobutyl adipate,diba,ftaflex diba,plasthall diba,bis 2-methylpropyl hexanedioate,hexanedioic acid, bis 2-methylpropyl ester,adipic acid, diisobutyl ester,diisobutyl hexanedioate,adipic acid bis 2-methylpropyl ester
8831
CHEBI:34708
bis(2-methylpropyl) hexanedioate
CC(C)COC(=O)CCCCC(=O)OCC(C)C
Flash Point >110°C (230°F)
Refractive Index 1.432
Density 0.954
Quantity 100g
Boiling Point 293°C
Formula Weight 258.36
Percent Purity 97%
Chemical Name or Material Diisobutyl adipate

EINECSNumber : 205-450-3

RTECSNumber : AV1480000

TSCA : Yes

Recommended Storage : Ambient temperatures