Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisobutylaluminium hydride, 1.1M solution in cyclohexane, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
1191-15-7, 110-82-7
C8H19Al
MFCD00008928
DIBAL-H, 1.1M solution in cyclohexane
1191-15-7, 110-82-7
C8H19Al
MFCD00008928
DIBAL-H, 1.1M solution in cyclohexane
Lewis acid, Reduction reagent
Linear Formula [(CH3)2CHCH2]2AIH
Beilstein 04, IV, 4400
Concentration 19.5 to 20.5% (DIBAL-H, on Al basis)
Fieser 01,260; 02,140; 03,101; 04,158; 05,224; 06,198; 07,111; 08,173; 09,171; 10,149; 11,185; 12,191; 13,115; 14,138; 15,137; 16,27; 17,123
Density 0.7800g/mL
Merck Index 15, 3212
Formula Weight 142.22
Physical Form Solution
Packaging AcroSeal™ Glass Bottle
Solubility Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic, hydrocarbons
Specific Gravity 0.78
Color Undesignated
Melting Point -70.0°C
Quantity 800mL
Flash Point −018°C
Chemical Name or Material Diisobutylaluminium hydride, 1.1M Solution in Cyclohexane
Show More Show Less

GHS H Statement:
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger