Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisobutylaluminium hydride, 1.2M (20 wt%) solution in toluene, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
400mL
800mL
Packaging:
AcroSeal™ Glass Bottle
1191-15-7,108-88-3
C8H19Al
142.22
MFCD00008928
DIBAL-H
1191-15-7,108-88-3
C8H19Al
142.22
MFCD00008928
DIBAL-H
Lewis acid, Reduction reagent
Density 0.8480g/mL
Packaging AcroSeal™ Glass Bottle
Quantity 800mL
Flash Point 4°C
Specific Gravity 0.848
Boiling Point 110.0°C
Color Undesignated
Linear Formula [(CH3)2CHCH2]2AIH
Beilstein 04, IV, 4400
Fieser 01,260; 02,140; 03,101; 04,158; 05,224; 06,198; 07,111; 08,173; 09,171; 10,149; 11,185; 12,191; 13,115; 14,138; 15,137; 16,27; 17,123
Merck Index 15, 3212
Solubility Information Solubility in water: reacts violently. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons
Formula Weight 142.22
Physical Form Solution
Chemical Name or Material Diisobutylaluminium hydride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May be fatal if swallowed and enters airways.
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging the unborn child.
May cause dro

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 214-729-9