Alfa Aesar™ Diisobutylaluminum chloride, 98+%, 25% w/w in hexane

Catalyst component in Ziegler-Natta type catalyst systems; in alkylation reactions

Manufacturer: Alfa Aesar™ 89039ND

Quantity 25g

 View more versions of this product

Catalog No. AA89039ND

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1779-25-5
    Molecular Formula C8H18AlCl
    Linear Formula [(CH3)2CHCH2]2AlCl
    Formula Weight 176.67
    MDL Number MFCD00008927
    UN Number UN3399
    Synonym DIBAC
    Chemical Name or Material Diisobutylaluminum chloride

    Specifications

    Formula Weight 176.67
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H225-H260-H304-H361-H373-H314-H336
    Highly flammable liquid and vapor.
    In contact with water releases flammable gases which may ignite spontaneously.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.
    DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
    Sensitivity Air & Moisture Sensitive
    Synonym DIBAC
    Health Hazard 1 Danger
    Physical Form Liquid
    Molecular Formula C8H18AlCl
    MDL Number MFCD00008927
    Quantity 25g
    UN Number UN3399
    Color Colorless
    Concentration 25% w/w in hexane
    TSCA TSCA
    Linear Formula [(CH3)2CHCH2]2AlCl
    CAS 1779-25-5
    Density 0.72g/mL
    Chemical Name or Material Diisobutylaluminum chloride
    Assay ≥98%

    GHS H Statement
    H225-H260-H304-H361-H373-H314-H336
    Highly flammable liquid and vapor.
    In contact with water releases flammable gases which may ignite spontaneously.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit