Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diisobutylaluminum hydride, 1M solution in hexane, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 42593AD

Quantity 50mL

 View more versions of this product

Catalog No. AA42593AD


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1191-15-7
142.22
UN3399
[(CH3)2CHCH2]2AlH
MFCD00008928
Diisobutylaluminum hydride

Specifications

1M solution in hexane
50mL
0.701g/mL
ChemSeal™ bottles
Liquid
142.22
UN3399
1191-15-7
−23°C (−10°F)
Colorless
Air & Moisture Sensitive
[(CH3)2CHCH2]2AlH
MFCD00008928
Diisobutylaluminum hydride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H260-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit