Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diisobutylaluminum hydride, 1M solution in toluene, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™
Click to view available options
Quantity:
50 mL
200 mL
500 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Packaged under Argon
Specifications
Color | Colorless |
Density | 0.805 g/mL |
Boiling Point | ∼110°C |
Flash Point | 4°C (40°F) |
Packaging | ChemSeal™ bottles |
Linear Formula | [(CH3)2CHCH2]2AlH |
Quantity | 50 mL |
UN Number | UN3399 |
Beilstein | 4123663 |
Sensitivity | Air and moisture sensitive |
Show More |
Safety and Handling
Hazard Category | H225-H250-H260-H304-H314-H335-H336-H361d-H373 |
Hazard Statement | GHS H Statement H224-H260-H304-H361-H373-H314-H318-H336 Extremely flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs (state all organs affected, if known) through prolonged or repeated exposure. Causes severe skin burns and eye damage. Causes serious eye damage. May cause drowsiness or dizziness. |
Precautionary Statement | P201-P202-P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE |
EINECSNumber | 214-729-9 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction