Alfa Aesar™ Diisobutylaluminum hydride, 25% w/w in hexane

Reducing agent

Manufacturer: Alfa Aesar™ 42588AD

Quantity 50mL

 View more versions of this product

Catalog No. AA42588AD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1191-15-7
    Linear Formula [(CH3)2CHCH2]2AlH
    Formula Weight 142.22
    MDL Number MFCD00008928
    UN Number UN3399
    Synonym DIBAL-H
    Chemical Name or Material Diisobutylaluminum hydride

    Specifications

    Formula Weight 142.22
    Health Hazard 2 GHS H Statement
    H224-H260-H304-H361-H373-H314-H318-H336
    Extremely flammable liquid and vapor.
    In contact with water releases flammable gases which may ignite spontaneously.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs (state all organs affected, if known) through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause drowsiness or dizziness.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Linear Formula [(CH3)2CHCH2]2AlH
    MDL Number MFCD00008928
    Sensitivity Air & Moisture Sensitive
    Health Hazard 1 Danger
    Density 0.701g/mL
    Color Colorless
    DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
    TSCA TSCA
    Physical Form Liquid
    CAS 1191-15-7
    Quantity 50mL
    UN Number UN3399
    Synonym DIBAL-H
    Concentration 25% w/w in hexane
    Chemical Name or Material Diisobutylaluminum hydride

    GHS H Statement
    H224-H260-H304-H361-H373-H314-H318-H336
    Extremely flammable liquid and vapor.
    In contact with water releases flammable gases which may ignite spontaneously.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs (state all organs affected, if known) through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit