Alfa Aesar™ Diisopentyl sulfide, 98%

Manufacturer: Alfa Aesar™ B2303006

Quantity 5g

 View more versions of this product

Catalog No. AAB2303006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 544-02-5
    Molecular Formula [(CH3)2CHCH2CH2]2S
    Formula Weight 174.35
    MDL Number MFCD00026522
    Synonym Bis(3-methylbutyl) sulfide; Isopentyl sulfide
    Chemical Name or Material Diisopentyl sulfide

    Specifications

    Formula Weight 174.35
    TSCA TSCA
    Boiling Point 79° to 80°C (7mmHg)
    Molecular Formula [(CH3)2CHCH2CH2]2S
    Quantity 5g
    Odor Unpleasant
    MDL Number MFCD00026522
    CAS 544-02-5
    Synonym Bis(3-methylbutyl) sulfide; Isopentyl sulfide
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Chemical Name or Material Diisopentyl sulfide
    Assay 98%

    Description

    Applications
    Diisopentyl sulfide is used as chemical, organic intermediates.

    Solubility
    Soluble in water (5.67mg/L @ 25°C.

    Notes
    Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit