Learn More
Diisopropanolamine, 98+%, sum of isomers, Thermo Scientific Chemicals
Description
Bis(2-hydroxypropyl)amine (Diisopropanolamine) is used to study its effects upon choline uptake and phospholipid synthesis in Chinese hamster ovary (CHO) cells. It is a versatile chemical that is widely used as an emulsifier, stabilizer, surfactant, and chemical building block. It can neutralize pH, act as a buffer (stabilize pH), or add basicity (alkalinity) to a solution. Major applications include personal care products, metalworking products, and natural gas purification. Diisopropanolamine is also used in coatings, paint strippers, paper, photographic intermediates, plastics, polyurethanes, and textiles. It is frequently used to neutralize fatty acids and sulfonic acidbased surfactants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Bis(2-hydroxypropyl)amine (Diisopropanolamine) is used to study its effects upon choline uptake and phospholipid synthesis in Chinese hamster ovary (CHO) cells. It is a versatile chemical that is widely used as an emulsifier, stabilizer, surfactant, and chemical building block. It can neutralize pH, act as a buffer (stabilize pH), or add basicity (alkalinity) to a solution. Major applications include personal care products, metalworking products, and natural gas purification. Diisopropanolamine is also used in coatings, paint strippers, paper, photographic intermediates, plastics, polyurethanes, and textiles. It is frequently used to neutralize fatty acids and sulfonic acidbased surfactants.
Solubility
Soluble in water 20°C 870g/L. Miscible in alcohol.
Notes
Air Sensitive & Hygroscopic. Store away from oxidizing agents, air, water/moisture and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
Melting Point | 38°C to 45°C |
Density | 1.004 |
Boiling Point | 249°C to 250°C |
Flash Point | 130°C (266°F) |
Odor | Amine-like |
Quantity | 250 g |
Beilstein | 605363 |
Sensitivity | Air sensitive; Hygroscopic |
Merck Index | 14,3195 |
Solubility Information | Soluble in water 20°C 870g/L. Miscible in alcohol. |
Show More |
Safety and Handling
Hazard Category | H319 |
Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
Precautionary Statement | P264b-P280i-P305+P351+P338 |
EINECSNumber | 203-820-9 |
RTECSNumber | UB6600000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.