Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropanolamine, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
40 lb.
Packaging:
Amber Glass Bottle
Poly Pail
110-97-4
110-97-4
Packaging Poly Pail
Quantity 40 lb.
CAS Max % 1
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.