Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diisopropyl azodicarboxylate, 94%, Thermo Scientific™

Click to view available options
Quantity:
25g
100g
2446-83-5
C8H14N2O4
202.21
MFCD00008875
VVWRJUBEIPHGQF-KTKRTIGZSA-N
z-n-propan-2-yloxy carbonyl imino propan-2-yloxy formamide
56776520
propan-2-yl (NZ)-N-propan-2-yloxycarbonyliminocarbamate
CC(C)OC(=O)N=NC(=O)OC(C)C
2446-83-5
C8H14N2O4
202.21
MFCD00008875
VVWRJUBEIPHGQF-KTKRTIGZSA-N
z-n-propan-2-yloxy carbonyl imino propan-2-yloxy formamide
56776520
propan-2-yl (NZ)-N-propan-2-yloxycarbonyliminocarbamate
CC(C)OC(=O)N=NC(=O)OC(C)C
Applications
Diisopropyl azodicarboxylate is used as an important reagent in the production of many organic compounds. It is used in association with triphenyl phosphine in Mitsunobu reaction of alcohols, acids and amides. It acts as reactant in the preparation of chromenes resembling classical cannabinoids, norbornene-based guanidine-rich polymers and acceptor-donor-acceptor organic dyes.

Solubility
Immiscible with water.

Notes
Store in cool place. Light sensitive. Incompatible with strong oxidizing agents, strong bases, alcohols and metallic salts. Decomposes violently at or above: 100 0C.
Sensitivity Light sensitive
Density 1.027
Refractive Index 1.42
Odor Pungent
Boiling Point 74°C to 75°C (0.3mmHg)
Flash Point 160°C (320°F)
Quantity 100g
Melting Point 3°C to 5°C
UN Number UN3082
Beilstein 1912326
Solubility Information Immiscible with water.
Formula Weight 202.21
Percent Purity 94%
Chemical Name or Material Diisopropyl azodicarboxylate
Show More Show Less

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P501c

H315-H319-H335-H373

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

EINECSNumber : 219-502-8

TSCA : No

Recommended Storage : Keep cold

RUO – Research Use Only