Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Diisopropyl D-tartrate, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
62961-64-2
C10H18O6
234.25
MFCD00008876
XEBCWEDRGPSHQH-YUMQZZPRSA-N
diisopropyl d-tartrate, 2s,3s-diisopropyl 2,3-dihydroxysuccinate, --diisopropyl d-tartrate, -diisopropyl-d-tartrate, d---tartaric acid diisopropyl ester, diisopropyl d---tartrate, butanedioic acid, 2,3-dihydroxy-, bis 1-methylethyl ester, 2s,3s, d---diisopropyl tartrate, d-dipt, --d-tartaric acid diisopropyl ester, diisoprropyl-d-tartrate
112972
dipropan-2-yl (2S,3S)-2,3-dihydroxybutanedioate
CC(C)OC(=O)C(C(C(=O)OC(C)C)O)O
62961-64-2
C10H18O6
234.25
MFCD00008876
XEBCWEDRGPSHQH-YUMQZZPRSA-N
diisopropyl d-tartrate, 2s,3s-diisopropyl 2,3-dihydroxysuccinate, --diisopropyl d-tartrate, -diisopropyl-d-tartrate, d---tartaric acid diisopropyl ester, diisopropyl d---tartrate, butanedioic acid, 2,3-dihydroxy-, bis 1-methylethyl ester, 2s,3s, d---diisopropyl tartrate, d-dipt, --d-tartaric acid diisopropyl ester, diisoprropyl-d-tartrate
112972
dipropan-2-yl (2S,3S)-2,3-dihydroxybutanedioate
CC(C)OC(=O)C(C(C(=O)OC(C)C)O)O
Flash Point 110°C
Specific Gravity 1.11
Boiling Point 275°C
Density 1.1100g/mL
Specific Rotation -17.25
Refractive Index 1.4380 to 1.44
Quantity 10g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless
Specific Rotation Condition −17.25° (20°C neat)
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula [-CH(OH)CO2CH(CH3)2]2
Fieser 13,276
Formula Weight 234.25
Physical Form Viscous Liquid
Percent Purity 99%
Chemical Name or Material (-)-Diisopropyl D-tartrate, 99%
Show More Show Less

HYGROSCOPIC

EINECSNumber : 263-771-4

TSCA : TSCA