Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Diisopropyl Ether, ReagentPlus™, 99%, Honeywell™

Click to view available options
Quantity:
1L
2.5L
4L
20L
Packaging:
Aluminum Bottle
Glass Bottle
Stainless Steel Can
108-20-3
C6H14O
102.177
MFCD00008880
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
diisopropyl ether,isopropyl ether,propane, 2,2'-oxybis,2-isopropoxypropane,diisopropyl oxide,diisopropylether,ether, isopropyl,2,2'-oxydipropane,isopropylether,bis isopropyl ether
7914
2-propan-2-yloxypropane
CC(C)OC(C)C
108-20-3
C6H14O
102.177
MFCD00008880
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
diisopropyl ether,isopropyl ether,propane, 2,2'-oxybis,2-isopropoxypropane,diisopropyl oxide,diisopropylether,ether, isopropyl,2,2'-oxydipropane,isopropylether,bis isopropyl ether
7914
2-propan-2-yloxypropane
CC(C)OC(C)C

Reagent Grade, contains either BHT or hydroquinone as stabilizer, 99%

  • ≥95% for general laboratory use.
Packaging Glass Bottle
Flash Point −29°C (−20.2°F)
Quantity 1L
Density 0.725g/mL at 25°C
Melting Point -86°C
Explosion Limit 1 to 21% (100°F)
Boiling Point 70°C
Refractive Index n20/D 1.367(lit.)
Vapor Density 3.5 (vs air)
Color Colorless
Vapor Pressure 120mmHg (20°C)
Ignition Point 827°F
Linear Formula (CH3)2CHOCH(CH3)2
UN Number UN1159
Beilstein 1731256
Formula Weight 102.17g/mol
Physical Form Liquid
Percent Purity 99%
Grade ReagentPlus™
Chemical Name or Material Diisopropyl Ether, Contains either BHT or hydroquinone as stabilizer
Show More Show Less

P210-P370 + P378-P403 + P235

H225-H336-H412

DOTInformation : Class 3 - PG 2 - Diisopropyl ether

EINECSNumber : 203-560-6

RTECSNumber : TZ5425000