Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (-)-Diisopropyl-L-malate, 97%

Click to view available options
Quantity:
5g
25g
100g
83541-68-8
C10H18O5
218.249
MFCD00274300
XYVHRFVONMVDGK-QMMMGPOBSA-N
diisopropyl s---malate,--diisopropyl-l-malate,1,4-diisopropyl 2s-2-hydroxybutanedioate,diisopropyl d-malate,s-diisopropyl malate,diisopropyl-s---malate,l-malic acid diisopropyl ester,s-2-hydroxybutanedioic acid diisopropyl ester
11333435
dipropan-2-yl (2S)-2-hydroxybutanedioate
CC(C)OC(=O)CC(C(=O)OC(C)C)O
83541-68-8
C10H18O5
218.249
MFCD00274300
XYVHRFVONMVDGK-QMMMGPOBSA-N
diisopropyl s---malate,--diisopropyl-l-malate,1,4-diisopropyl 2s-2-hydroxybutanedioate,diisopropyl d-malate,s-diisopropyl malate,diisopropyl-s---malate,l-malic acid diisopropyl ester,s-2-hydroxybutanedioic acid diisopropyl ester
11333435
dipropan-2-yl (2S)-2-hydroxybutanedioate
CC(C)OC(=O)CC(C(=O)OC(C)C)O
Quantity 25g
Boiling Point 236°C to 238°C
Formula Weight 218.25
Percent Purity 97%
Chemical Name or Material (-)-Diisopropyl-L-malate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning