Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Diisopropyl L-tartrate, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
2217-15-4
C10H18O6
234.248
MFCD00064450
XEBCWEDRGPSHQH-HTQZYQBOSA-N
+-diisopropyl l-tartrate,2r,3r-diisopropyl 2,3-dihydroxysuccinate,di-isopropyl tartrate,diisoprropyl-l-tartrate,+-dipt,l-+-tartaric acid diisopropyl ester,diisopropyl l-tartrate,l-+-diisopropyl-tartrate, l-dipt,butanedioic acid, 2,3-dihydroxy-, bis 1-methylethyl ester, r,r-+/-,1,4-diisopropyl 2r,3r-2,3-dihydroxybutanedioate
6453580
dipropan-2-yl (2R,3R)-2,3-dihydroxybutanedioate
CC(C)OC(=O)C(C(C(=O)OC(C)C)O)O
2217-15-4
C10H18O6
234.248
MFCD00064450
XEBCWEDRGPSHQH-HTQZYQBOSA-N
+-diisopropyl l-tartrate,2r,3r-diisopropyl 2,3-dihydroxysuccinate,di-isopropyl tartrate,diisoprropyl-l-tartrate,+-dipt,l-+-tartaric acid diisopropyl ester,diisopropyl l-tartrate,l-+-diisopropyl-tartrate, l-dipt,butanedioic acid, 2,3-dihydroxy-, bis 1-methylethyl ester, r,r-+/-,1,4-diisopropyl 2r,3r-2,3-dihydroxybutanedioate
6453580
dipropan-2-yl (2R,3R)-2,3-dihydroxybutanedioate
CC(C)OC(=O)C(C(C(=O)OC(C)C)O)O
Infrared Spectrum Authentic
Quantity 100g
Refractive Index 1.4380 to 1.4400 (20°C, 589nm)
Color Colorless
Density 1.11g/mL
Specific Rotation Condition +17.25° (20°C neat)
Packaging Glass bottle
Specific Rotation +16° to +18.5° (20°C, 589nm) (neat)
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula {-CH(OH)CO2CH(CH3)2}2
Beilstein 03, 517
Fieser 13,276; 16,339; 12,493; 13,310
Formula Weight 234.25
Physical Form Viscous Liquid
Percent Purity ≥98.5%
Chemical Name or Material (+)-Diisopropyl L-tartrate, 99%
Show More Show Less

HYGROSCOPIC

EINECSNumber : 218-709-0

RTECSNumber : WW8100000

TSCA : TSCA