Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyldichlorosilane, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
7751-38-4
C6H14Cl2Si
185.17
GSENNYNYEKCQGA-UHFFFAOYSA-N
diisopropyldichlorosilane,dichlorodiisopropylsilane,silane, dichlorobis 1-methylethyl,dichloro-di propan-2-yl silane,silane, dichlorodiisopropyl,acmc-209pad,dichloro diisopropyl silane,di 2-propyl dichlorosilane,dichlorobis propan-2-yl silane,bis chloranyl-di propan-2-yl silane
2758384
dichloro-di(propan-2-yl)silane
CC(C)[Si](C(C)C)(Cl)Cl
7751-38-4
C6H14Cl2Si
185.17
GSENNYNYEKCQGA-UHFFFAOYSA-N
diisopropyldichlorosilane,dichlorodiisopropylsilane,silane, dichlorobis 1-methylethyl,dichloro-di propan-2-yl silane,silane, dichlorodiisopropyl,acmc-209pad,dichloro diisopropyl silane,di 2-propyl dichlorosilane,dichlorobis propan-2-yl silane,bis chloranyl-di propan-2-yl silane
2758384
dichloro-di(propan-2-yl)silane
CC(C)[Si](C(C)C)(Cl)Cl
Specific Gravity 1.026
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point 43°C
Density 1.0260g/mL
Quantity 25g
Color Colorless
Boiling Point 168.0°C to 170.0°C
Refractive Index 1.4430 to 1.4450 (20°C, 589nm)
Assay Percent Range 96% min. (GC)
Formula Weight 185.17
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Diisopropyldichlorosilane
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Contact with water liberates toxic gas.

GHS P Statement
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.

GHS Signal Word: Danger