Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropylformamide, 98+%, ACROS Organics™

Click to view available options
Quantity:
5mL
Packaging:
Glass bottle
2700-30-3
C7H15NO
129.203
MFCD00008867
UNBDDZDKBWPHAX-UHFFFAOYSA-N
n,n-diisopropylformamide,formamide, n,n-bis 1-methylethyl,n-formyldiisopropylamine,diisopropylformamide,n,n-bis 1-methylethyl formamide,nn-di-iso-propylformamide,formamide, n,n-diisopropyl,formamide, bis 1-methylethyl,n,n-bis methylethyl carboxamide,1y1&nvhy1&1
75912
N,N-di(propan-2-yl)formamide
CC(C)N(C=O)C(C)C
2700-30-3
C7H15NO
129.203
MFCD00008867
UNBDDZDKBWPHAX-UHFFFAOYSA-N
n,n-diisopropylformamide,formamide, n,n-bis 1-methylethyl,n-formyldiisopropylamine,diisopropylformamide,n,n-bis 1-methylethyl formamide,nn-di-iso-propylformamide,formamide, n,n-diisopropyl,formamide, bis 1-methylethyl,n,n-bis methylethyl carboxamide,1y1&nvhy1&1
75912
N,N-di(propan-2-yl)formamide
CC(C)N(C=O)C(C)C
Refractive Index 1.4360 to 1.4380 (20°C, 589nm)
Packaging Glass bottle
Density 0.8900g/mL
Infrared Spectrum Authentic
Quantity 5mL
Color Brown
Assay Percent Range 98+%
Linear Formula HCON[CH(CH3)2]2
Beilstein 04, IV, 515
Fieser 08,174
Formula Weight 129.2
Physical Form Liquid
Percent Purity ≥98.0%
Chemical Name or Material Diisopropylformamide, 98+%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

COMBUSTIBLE LIQUID