Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropylphosphine, 10 wt.% solution in hexanes, Acros Organics

Click to view available options
Quantity:
2.5g
10g
50g
Packaging:
Glass bottle
20491-53-6,110-54-3
C6H15P
118.16
MFCD00233814
WDIIYWASEVHBBT-UHFFFAOYSA-N
diisopropylphosphine,di-isopropylphosphine,di-i-propylphosphine,diisopropylphosphane,phosphine, diisopropyl,phosphine, bis 1-methylethyl,ipr2ph,di-iso-propylphosphine,di-iso-propyl phosphine,di propan-2-yl phosphane
536496
di(propan-2-yl)phosphane
CC(C)PC(C)C
20491-53-6,110-54-3
C6H15P
118.16
MFCD00233814
WDIIYWASEVHBBT-UHFFFAOYSA-N
diisopropylphosphine,di-isopropylphosphine,di-i-propylphosphine,diisopropylphosphane,phosphine, diisopropyl,phosphine, bis 1-methylethyl,ipr2ph,di-iso-propylphosphine,di-iso-propyl phosphine,di propan-2-yl phosphane
536496
di(propan-2-yl)phosphane
CC(C)PC(C)C
Specific Gravity 0.67
Quantity 2.5g
Flash Point −22°C
Density 0.6700g/mL
NMR complies (31P-NMR)
Color Brown-Yellow to Yellow
Packaging Glass bottle
Linear Formula [(CH3)2CH]2PH
Solubility Information Solubility in water: insoluble
Formula Weight 118.16
Physical Form Liquid
Percent Purity 9 to 11% (Total base)
Chemical Name or Material Diisopropylphosphine
Show More Show Less

GHS H Statement
May be fatal if swallowed and enters airways. Causes severe skin burns and eye damage. May cause damage to organs through prolonged or repeated exposure. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Highly flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

GHS Signal Word: Danger