Learn More
Sklar InstrumentsSupplier Diversity Partner Walther Dilator-Catheter, Sklar, 5-1/2 in., Curved, Chrome Plated Steel. PK/14. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 852010
Walther Dilator-Catheter, Sklar, 5-1/2 in., Curved, Chrome Plated Steel. Sklar's Walther Dilator-Catheter is a female urethral sound commonly used to dilate the female urethra to ease placement of a cystoscope. It may also be used to take a urine specimen or drain the bladder. This product is a set of 14 devices, size 12 to 38 French.
Catalog No. 50-143-6398