Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dilauroyl peroxide, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
105-74-8
C24H46O4
398.63
MFCD00008964
YIVJZNGAASQVEM-UHFFFAOYSA-N
lauroyl peroxide,dilauroyl peroxide,dodecanoyl peroxide,peroxide, bis 1-oxododecyl,laurydol,laurox,alperox c,dodecanoic peroxyanhydride,dyp-97f,peroxyde de lauroyle
7773
CHEBI:82479
dodecanoyl dodecaneperoxoate
CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC
105-74-8
C24H46O4
398.63
MFCD00008964
YIVJZNGAASQVEM-UHFFFAOYSA-N
lauroyl peroxide,dilauroyl peroxide,dodecanoyl peroxide,peroxide, bis 1-oxododecyl,laurydol,laurox,alperox c,dodecanoic peroxyanhydride,dyp-97f,peroxyde de lauroyle
7773
CHEBI:82479
dodecanoyl dodecaneperoxoate
CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC
Radical initiators
Quantity 500g
Infrared Spectrum Authentic
Packaging Plastic bottle
Melting Point 53°C to 57°C
Color White
Assay Percent Range 98.5% min. (Iodometry)
Linear Formula [CH3(CH2)10CO]2O2
Solubility Information Solubility in water: insoluble. Other solubilities: partly soluble in toluene and hexane
Formula Weight 398.63
Physical Form Powder
Percent Purity 99%
Chemical Name or Material Dilauroyl peroxide, 99%
Show More Show Less

GHS H Statement
Heating may cause a fire.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/ combustible materials.
Keep only in orig

GHS Signal Word: Danger

EINECSNumber : 203-326-3

RTECSNumber : OF2625000

TSCA : TSCA