Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diltiazem Hydrochloride, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
33286-22-5
C22H27ClN2O4S
450.98
HDRXZJPWHTXQRI-BHDTVMLSSA-N
hydrogen (2S,3S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl acetate chloride
[H+].[Cl-].COC1=CC=C(C=C1)[C@@H]1SC2=CC=CC=C2N(CCN(C)C)C(=O)[C@@H]1OC(C)=O
33286-22-5
C22H27ClN2O4S
450.98
HDRXZJPWHTXQRI-BHDTVMLSSA-N
hydrogen (2S,3S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl acetate chloride
[H+].[Cl-].COC1=CC=C(C=C1)[C@@H]1SC2=CC=CC=C2N(CCN(C)C)C(=O)[C@@H]1OC(C)=O

D1163, 33286-22-5, C22H26N2O4S•HCl

Spectrum™ Diltiazem Hydrochloride, USP is a calcium channel blocker used to prevent chest pain (angina). All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Pail
Ignition Residue 0.001
Loss on Drying 0.005
Quantity 5 kg
Specific Rotation +110° to +116°
CAS Max % 1
Percent Purity 98 to 102%