Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIM, Tocris Bioscience™

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
C17H14N2
3,3'-diindolylmethane,di 1h-indol-3-yl methane,diindolylmethane,3,3'-methylenediindole,3,3'-methylenebis-1h-indole,dim,1h-indole, 3,3'-methylenebis,3,3'-di-indolylmethane,3-diindolyl methane,arundine
DIM
This item is not returnable. View return policy
C17H14N2
3,3'-diindolylmethane,di 1h-indol-3-yl methane,diindolylmethane,3,3'-methylenediindole,3,3'-methylenebis-1h-indole,dim,1h-indole, 3,3'-methylenebis,3,3'-di-indolylmethane,3-diindolyl methane,arundine
DIM

Inhibits EGFR activation; also induces apoptosis

Activator of Chk2 that causes G2/M cell cycle arrest in various cancer cell lines. Antiproliferative and inducer of apoptosis; promotes proteasomal degradation of Cdc25C and Cdk1. Inhibits phosphorylation of EGFR and downstream activation of ERK.
Quantity 50mg
CAS 1968-05-4
InChI Key VFTRKSBEFQDZKX-UHFFFAOYSA-N
Molecular Weight (g/mol) 246.31
PubChem CID 3071
ChEBI CHEBI:50182
Formula Weight 246.31

Recommended Storage : Store at + 4°C