Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimebon dihydrochloride, Tocris Bioscience™

Catalog Number 32011
Click to view available options
Quantity:
1 mg
10 mg
This item is not returnable. View return policy
97657-92-6
C21H27Cl2N3
392.368
GTWLIQOLGOZTLF-UHFFFAOYSA-N
dimebon dihydrochloride, dimebolin dihydrochloride, latrepirdine dihydrochloride, unii-8b9414qq5m, latrepirdine dihydrochloride usan, 2,8-dimethyl-5-2-6-methylpyridin-3-yl ethyl-2,3,4,5-tetrahydro-1h-pyrido 4,3-b indole dihydrochloride, dimebone dihydrochloride hydrate, dimebolin dihydrochloride hydrate
23729232
2,8-dimethyl-5-[2-(6-methylpyridin-3-yl)ethyl]-3,4-dihydro-1H-pyrido[4,3-b]indole;dihydrochloride
CC1=CC2=C(C=C1)N(C3=C2CN(CC3)C)CCC4=CN=C(C=C4)C.Cl.Cl
This item is not returnable. View return policy
97657-92-6
C21H27Cl2N3
392.368
GTWLIQOLGOZTLF-UHFFFAOYSA-N
dimebon dihydrochloride, dimebolin dihydrochloride, latrepirdine dihydrochloride, unii-8b9414qq5m, latrepirdine dihydrochloride usan, 2,8-dimethyl-5-2-6-methylpyridin-3-yl ethyl-2,3,4,5-tetrahydro-1h-pyrido 4,3-b indole dihydrochloride, dimebone dihydrochloride hydrate, dimebolin dihydrochloride hydrate
23729232
2,8-dimethyl-5-[2-(6-methylpyridin-3-yl)ethyl]-3,4-dihydro-1H-pyrido[4,3-b]indole;dihydrochloride
CC1=CC2=C(C=C1)N(C3=C2CN(CC3)C)CCC4=CN=C(C=C4)C.Cl.Cl

Neuroprotectant; protects against β-amyloid neurotoxicity

Non-selective antihistamine that displays cognitive enhancing abilities. Also displays high affinity for 5-HT (particularly 5-HT6 and 5-HT7), α-adrenergic, dopaminergic, AMPA and NMDA receptors, and L-type calcium channels. Does not inhibit acetylcholinesterase activity. Exhibits neuroprotective activity in cellular models of Alzheimer's and Huntington's disease and preserves cognitive function following administration to AF64A lesioned rats. Protects neurons against the neurotoxic action of β-amyloid fragment; shown to enhance autophagy in yeast and reduce intracellular Aβ42 levels.
Quantity 1 mg
Formula Weight 392.37
Percent Purity >98%
Chemical Name or Material Dimebon dihydrochloride
Recommended Storage Desiccate at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit